MilliporeSigma
  • Presynaptic effect of L-glutamic acid on the release of dopamine in rat striatal slices.
Presynaptic effect of L-glutamic acid on the release of dopamine in rat striatal slices.
Neuroscience letters (1977-10-01)
M F Giorguieff, M L Kemel, J Glowinski