MilliporeSigma
  • Heterogeneous palladium-catalysed heck reaction of aryl chlorides and styrene in water under mild conditions.
Heterogeneous palladium-catalysed heck reaction of aryl chlorides and styrene in water under mild conditions.
Advanced Synthesis & Catalysis (2002)
Mukhopadhyay S, et al.