MilliporeSigma
  • Chlorinated Decreasing Solvents: Physical-Chemical Properties Affecting Aquifer Contamination and Remediation.
Chlorinated Decreasing Solvents: Physical-Chemical Properties Affecting Aquifer Contamination and Remediation.
Ground Water Monitoring & Remediation (1999)
Jackson RE and Dwarakanath V.