MilliporeSigma
  • Clean synthesis of adipic acid by direct oxidation of cyclohexene with H2O2 over peroxytungstate-organic complex catalysts.
Clean synthesis of adipic acid by direct oxidation of cyclohexene with H<SUB>2</SUB>O<SUB>2</SUB> over peroxytungstate-organic complex catalysts.
Green Chemistry (1999)
Deng Y, et al.