- Cerium (III) chloride promoted highly regioselective ring opening of epoxides and aziridines using NaN3 in acetonitrile: a facile synthesis of 1,2-azidoalcohols and 1,2-azidoamines.
Cerium (III) chloride promoted highly regioselective ring opening of epoxides and aziridines using NaN<SUB>3</SUB> in acetonitrile: a facile synthesis of 1,2-azidoalcohols and 1,2-azidoamines.
Organic Letters (2002)
Sabitha G, et al.