- Competitive Formation of Endoperoxide and Hydroperoxide in the Reaction of Singlet Oxygen with α-Terpinene.
Competitive Formation of Endoperoxide and Hydroperoxide in the Reaction of Singlet Oxygen with α-Terpinene.
ChemInform (1991)
Matusch R and Schmidt G.