MilliporeSigma
  • Nickel-catalyzed regio- and stereoselective silylation of terminal alkenes with silacyclobutanes: facile access to vinylsilanes from alkenes.
Nickel-catalyzed regio- and stereoselective silylation of terminal alkenes with silacyclobutanes: facile access to vinylsilanes from alkenes.
Journal of the American Chemical Society (2007-04-20)
Koji Hirano, Hideki Yorimitsu, Koichiro Oshima
Sigma-Aldrich
2-Vinylpyridine, 97%