MilliporeSigma
  • Studies on the composition of glyceryl ethers and their preparation from diacyl glyceryl ethers in liver oils.
Studies on the composition of glyceryl ethers and their preparation from diacyl glyceryl ethers in liver oils.
Journal of Lipid Research (1963)
Guyer KE, et al.