MilliporeSigma
  • A protocol for preparing, characterizing and using three RNA-specific, live cell imaging probes: E36, E144 and F22
A protocol for preparing, characterizing and using three RNA-specific, live cell imaging probes: E36, E144 and F22
Nature Protocols (2006)
Li Q & Chang YT