MilliporeSigma
  • Bis (diphenylphosphino) acetonitrile: Synthesis, ligand properties and application in catalytic carbon?carbon coupling
Bis (diphenylphosphino) acetonitrile: Synthesis, ligand properties and application in catalytic carbon?carbon coupling
Dalton Transactions (2007)
Braun L, et al.