MilliporeSigma
  • Cobalt-catalysed three-component coupling involving ketones or ketoesters, aldehydes and acetonitrile: a novel one-pot synthesis of ?-acetamido ketones
Cobalt-catalysed three-component coupling involving ketones or ketoesters, aldehydes and acetonitrile: a novel one-pot synthesis of ?-acetamido ketones
Journal of the Chemical Society. Chemical Communications (1994)
Madhavaa Reddy M.