MilliporeSigma
  • Catalytic performance of metal oxides for the synthesis of propylene carbonate from urea and 1, 2-propanediol.
Catalytic performance of metal oxides for the synthesis of propylene carbonate from urea and 1, 2-propanediol.
J. Mol. Catal. A: Chem. (2007)
Li Q, et al.