- Epoxidation of cyclooctene and cyclohexene with hydrogen peroxide catalyzed by bis [3, 5-bis (trifluoromethyl)-diphenyl] diselenide: Recyclable catalyst-containing phases through the use of glycerol-derived solvents
Epoxidation of cyclooctene and cyclohexene with hydrogen peroxide catalyzed by bis [3, 5-bis (trifluoromethyl)-diphenyl] diselenide: Recyclable catalyst-containing phases through the use of glycerol-derived solvents
J. Mol. Catal. A: Chem. (2011)
Garcia-Marin H, et al.