MilliporeSigma
  • An endophytic Nodulisporium sp. producing volatile organic compounds having bioactivity and fuel potential.
An endophytic Nodulisporium sp. producing volatile organic compounds having bioactivity and fuel potential.
Journal of Petroleum & Environmental Biotechnology (2012)
Mends MT, et al.