MilliporeSigma
  • A comparison of essential oil constituents of bark, leaf, root and fruit of cinnamon (Cinnamomum zeylanicum Blum) grown in Sri Lanka.
A comparison of essential oil constituents of bark, leaf, root and fruit of cinnamon (Cinnamomum zeylanicum Blum) grown in Sri Lanka.
Journal of the National Science Foundation of Sri Lanka (2010)
Paranagama PA, et al.