MilliporeSigma
  • Extraction of essential oils from five cinnamon leaves and identification of their volatile compound compositions.
Extraction of essential oils from five cinnamon leaves and identification of their volatile compound compositions.
Innovative Food Science & Emerging Technologies (2009)
Wang R, et al.