MilliporeSigma
Sensor Dyes
Monika Baeumle
BioFiles 2011, 6.3, 23.
Nitric Oxide Probes
In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide (NO) as a signaling molecule in the cardiovascular system. This indicates the importance the scientific community has placed on the role of nitric oxide as a signal transporter in neurons, endothelial cells, and in the immune system. NO has been implicated in vasodilation,1 neurotransmission,2 cytotoxicity, immune response,3 and inflammation.4-6
Within cells, nitric oxide synthase (NOS) catalyzes the conversion of arginine to citrulline plus NO in the presence of molecular oxygen, tetrahydrobiopterin, NADPH, and flavin cofactors. Nε-hydroxy-L-arginine is an intermediate in this reaction. NO has an in vivo half-life of only a few seconds;7 however, since it is soluble in both aqueous and lipophilic media, it readily diffuses through the cytoplasm and across cell membranes. NO has effects on neuronal transmission as well as synaptic plasticity in the central nervous system. NO activates guanylate cyclase in the vasculature, thus, increasing the synthesis of cGMP, which in turn induces smooth muscle relaxation and vasodilation. NO toxicity is related to its ability to combine with superoxide anions to form peroxynitrite, an oxidizing free radical that can damage DNA and other cellular constituents.
The NO molecule is unstable and occurs in very low concentrations in biological systems. In order to detect it in real time, methods with high sensitivities and specificities are required. Fluorescence microscopy seems to be the method of choice, provided suitable NO probes are available. This high spatial-temporal resolution technique allows for reliable imaging of in situ NO concentrations.
Research into the design of sensitive NO probes revealed a reaction involving vicinal aromatic diamino compounds8,9 that allow for selective trapping and detection of NO. Figure 1 illustrates this reaction using the novel NO probe 5,6-diaminofluorescein diacetate (DAF-2-DA) as an example.
Dye figure 1
Figure 1.Principle of intracellular NO measurement, shown with the probe DAF-2 DA[10]. Lipophilic, non-fluorescent DAF-2-DA permeates the cell membrane and is then hydrolyzed by cytosolic esterases to the weakly fluorescent probe DAF-2. The presence of NO radicals transforms it to the strongly fluorescent triazole DAF-2 T.
Naphthalenes
2,3-Diaminonaphthalene (DAN) was established as a sensitive probe to measure NO-concentration in vitro and under physiological conditions.11 The respective 2,3-naphthotriazole (NAT) was detected in the low nM concentration range. DAN, however, proved to be ill-suited to NO detection in cells due to rapid leakage through cell membranes after loading. High background fluorescence in biological matrices was also experienced due to its rather short excitation and emission wavelengths.
Therefore, DAN-1 and its cell membrane permeable precursor DAN-1-EE were developed.12DAN-1 is formed by the hydrolysis of DAN-1-EE by cytosolic esterases. DAN-1-T (Figure 2), the fluorescent reaction product of DAN-1 with NO, showed considerably improved leakage behavior.
Naphthalenes
Figure 2.Time course of formation of fluorescent DAN-1-T from DAN-1 upon addition of final concentration of 5 mM (green line) or 50 mM (red line) respectively of a NO forming agent (NONOate)[12].
Xanthenes
Fluorescein and rhodamine-based probes have been developed in order to eliminate some drawbacks of naphthalene-based NO probes, such as cytotoxicity, strong autofluorescence, a small extinction coefficient, poor solubility in neutral buffer, and short excitation/emisson wavelengths.10,13-16
Limit of detection (LOD) of NO by DAF-2 is 5 nM in vitro, in the absence of absorbing side products. Oxidized forms of NO such as NO2- and NO3-, as well as reactive oxygen species such as O2 ·-,H2O2, and ONOO- do not react with DAF-2 to give a fluorescent product. Therefore, under physiological conditions fluorescent DAF-2 T is only formed in the presence of NO.
A fluorescent response was detected when cultured, smooth aortic rat muscle cells loaded with DAF-2-DA were viewed using a confocal laser scanning microscope in the presence of endotoxins and cytokines. The addition of L-arginine resulted in a sharp increase of the fluorescence signal, whereas supplementary addition of N-monomethyl-L-arginine (L-NMMA) resulted in no signal increase, clearly demonstating the inhibition of NO-synthase.
The fluorescein type probe (DAF-2) shows a bright fluorescence signal, while the rhodamine type probes (DAR-1 and DAR-2) excel with high photostability and their applicability over a broad pH range (Table 1, Table 2 and Figure 3).
probeλab1, max. [nm]
∈ [x10-4 M-1cm-1]
λcm, max. [nm]
rel.quantum yields
 diaminetriazolediaminetriazoletriazolediaminetriazole
DAF-24864917.97.35130.0050.92
DAR-1550556128.75750.0040.25
DAR-2549552107.15710.0060.34
Table 1Spectral properties of DAF and DAR probesa)b)
  1. Data from reference.11
  2. Data at 20 °C in 0.1 M phosphate buffer, pH 7.4; quantum yields derived by comparison with known quantum yield of rhodamin B in ethanol.
 DAF'sDAR's
λcx [nm]-490-550
λcm [nm]-515 (green)-575 (red)
suitable pH range>pH6>pH4
detection limit for No-5 nM (DAF-2, in vitro)-10 nM (DAR-M, in vitro)
photostabilityb
photolbeached more than 70%not photobleached
distribution nucleus/cytosolequalcytosol>nucleus
Table 2General Properties of DAF and DAR probes
  1. Data from reference.10-12
  2. Based on fluorescence intensity measurements after exposure to sunlight for 3 h.
pH dependence of fluorescence intensity
Figure 3.pH dependence of fluorescence intensity (a.u.) of DAF-2 (green), DAR-1 (orange) and DAR-2 (pink)[11].
Oxygen Probes
The detection of molecular oxygen plays an important role in many applications and biological processes. The probes are oxygen-sensitive indicators that change their fluorescense intensity or emission maximum after binding to molecular oxygen. The sensors are reversible and are stable against photobleaching.
ROS Probes
Reactive oxygen probes (ROS) are associated with research in many diseases like inflammatory and cancer, where increased levels of ROS are produced in cells. The study of ROS requires photostable sensors with minimal autooxidation and low production rates of by-products.17
Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
References
1.
Palmer RMJ, Ferrige AG, Moncada S. 1987. Nitric oxide release accounts for the biological activity of endothelium-derived relaxing factor. Nature. 327(6122):524-526. https://doi.org/10.1038/327524a0
2.
Garthwaite J, Charles SL, Chess-Williams R. 1988. Endothelium-derived relaxing factor release on activation of NMDA receptors suggests role as intercellular messenger in the brain. Nature. 336(6197):385-388. https://doi.org/10.1038/336385a0
3.
Marletta MA, Yoon PS, Iyengar R, Leaf CD, Wishnok JS. 1988. Macrophage oxidation of L-arginine to nitrite and nitrate: nitric oxide is an intermediate. Biochemistry. 27(24):8706-8711. https://doi.org/10.1021/bi00424a003
4.
Stuehr DJ. 1997. STRUCTURE-FUNCTION ASPECTS IN THE NITRIC OXIDE SYNTHASES. Annu. Rev. Pharmacol. Toxicol.. 37(1):339-359. https://doi.org/10.1146/annurev.pharmtox.37.1.339
5.
Marletta MA, Hurshman AR, Rusche KM. 1998. Catalysis by nitric oxide synthase. Current Opinion in Chemical Biology. 2(5):656-663. https://doi.org/10.1016/s1367-5931(98)80098-7
6.
Geller DA, Billiar TR. 1998. 17(1):7-23. https://doi.org/10.1023/a:1005940202801
7.
Mordvintcev P, Mülsch A, Busse R, Vanin A. 1991. On-line detection of nitric oxide formation in liquid aqueous phase by electron paramagnetic resonance spectroscopy. Analytical Biochemistry. 199(1):142-146. https://doi.org/10.1016/0003-2697(91)90282-x
8.
Misko T, Schilling R, Salvemini D, Moore W, Currie M. 1993. A Fluorometric Assay for the Measurement of Nitrite in Biological Samples. Analytical Biochemistry. 214(1):11-16. https://doi.org/10.1006/abio.1993.1449
9.
Miles AM, Wink DA, Cook JC, Grisham MB. 1996. Determination of nitric oxide using fluorescence spectroscopy.105-120. https://doi.org/10.1016/s0076-6879(96)68013-6
10.
Kojima H, Nakatsubo N, Kikuchi K, Kawahara S, Kirino Y, Nagoshi H, Hirata Y, Nagano T. 1998. Detection and Imaging of Nitric Oxide with Novel Fluorescent Indicators:  Diaminofluoresceins. Anal. Chem.. 70(13):2446-2453. https://doi.org/10.1021/ac9801723
11.
Andrew PJ, Auer M, Lindley IJ, Kauffmann HF, Kungl AJ. 1997. Intensity-independent fluorometric detection of cellular nitric oxide release. 408(3):319-323. https://doi.org/10.1016/s0014-5793(97)00449-3
12.
KOJIMA H, SAKURAI K, KIKUCHI K, KAWAHARA S, KIRINO Y, NAGOSHI H, HIRATA Y, AKAIKE T, MAEDA H, NAGANO T. 1997. Development of a Fluorescent Indicator for the Bioimaging of Nitric Oxide.. Biol. Pharm. Bull.. 20(12):1229-1232. https://doi.org/10.1248/bpb.20.1229
13.
Kojima H, Hirotani M, Urano Y, Kikuchi K, Higuchi T, Nagano T. 2000. Fluorescent indicators for nitric oxide based on rhodamine chromophore. Tetrahedron Letters. 41(1):69-72. https://doi.org/10.1016/s0040-4039(99)02002-x
14.
Kojima H, Hirotani M, Nakatsubo N, Kikuchi K, Urano Y, Higuchi T, Hirata Y, Nagano T. 2001. Bioimaging of Nitric Oxide with Fluorescent Indicators Based on the Rhodamine Chromophore. Anal. Chem.. 73(9):1967-1973. https://doi.org/10.1021/ac001136i
15.
KOJIMA H, SAKURAI K, KIKUCHI K, KAWAHARA S, KIRINO Y, NAGOSHI H, HIRATA Y, NAGANO T. 1998. Development of a Fluorescent Indicator for Nitric Oxide Based on the Fluorescein Chromophore.. Chem. Pharm. Bull.. 46(2):373-375. https://doi.org/10.1248/cpb.46.373
16.
Nakatsubo N, Kojima H, Kikuchi K, Nagoshi H, Hirata Y, Maeda D, Imai Y, Irimura T, Nagano T. 1998. Direct evidence of nitric oxide production from bovine aortic endothelial cells using new fluorescence indicators: diaminofluoresceins. 427(2):263-266. https://doi.org/10.1016/s0014-5793(98)00440-2
17.
Xu H, Aylott JW, Kopelman R, Miller TJ, Philbert MA. 2001. A Real-Time Ratiometric Method for the Determination of Molecular Oxygen Inside Living Cells Using Sol?Gel-Based Spherical Optical Nanosensors with Applications to Rat C6 Glioma. Anal. Chem.. 73(17):4124-4133. https://doi.org/10.1021/ac0102718