Merck
  • RNA Helicase DDX1 Converts RNA G-Quadruplex Structures into R-Loops to Promote IgH Class Switch Recombination.
RNA Helicase DDX1 Converts RNA G-Quadruplex Structures into R-Loops to Promote IgH Class Switch Recombination.
Molecular cell (2018-05-08)
Claudia Ribeiro de Almeida, Somdutta Dhir, Ashish Dhir, Amin E Moghaddam, Quentin Sattentau, Anton Meinhart, Nicholas J Proudfoot
ZUSAMMENFASSUNG
Class switch recombination (CSR) at the immunoglobulin heavy-chain (IgH) locus is associated with the formation of R-loop structures over switch (S) regions. While these often occur co-transcriptionally between nascent RNA and template DNA, we now show that they also form as part of a post-transcriptional mechanism targeting AID to IgH S-regions. This depends on the RNA helicase DDX1 that is also required for CSR in vivo. DDX1 binds to G-quadruplex (G4) structures present in intronic switch transcripts and converts them into S-region R-loops. This in turn targets the cytidine deaminase enzyme AID to S-regions so promoting CSR. Notably R-loop levels over S-regions are diminished by chemical stabilization of G4 RNA or by the expression of a DDX1 ATPase-deficient mutant that acts as a dominant-negative protein to reduce CSR efficiency. In effect, we provide evidence for how S-region transcripts interconvert between G4 and R-loop structures to promote CSR in the IgH locus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Freund-Adjuvans, komplett, cell suspension
Sigma-Aldrich
Puromycin -dihydrochlorid aus Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Freund-Adjuvans, inkomplett, liquid
Sigma-Aldrich
Monoklonale ANTI-FLAG® M2-Peroxidase (HRP) in Maus hergestellte Antikörper, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
TAT-CRE-Recombinase, TAT-CRE Recombinase is a recombinant cell-permeant fusion cre-recombinase protein consisting of TAT sequence, a nuclear localization sequence (NLS) and it is known to catalyze the site specific recombination event between two loxP DNA sites.
Sigma-Aldrich
5,6-Dichlorbenzimidazol 1-β-D-Ribofuranosid
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
Pyridostatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Anti-CTCF-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Mouse IgM (μ-chain specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Pyridostatin trifluoroacetate salt, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ddx1
Sigma-Aldrich
MISSION® esiRNA, targeting human DDX1