Merck
  • Quantitative phosphoproteomic analysis of T cell receptor signaling reveals system-wide modulation of protein-protein interactions.
Quantitative phosphoproteomic analysis of T cell receptor signaling reveals system-wide modulation of protein-protein interactions.
Science signaling (2009-08-20)
Viveka Mayya, Deborah H Lundgren, Sun-Il Hwang, Karim Rezaul, Linfeng Wu, Jimmy K Eng, Vladimir Rodionov, David K Han
ZUSAMMENFASSUNG
Protein phosphorylation events during T cell receptor (TCR) signaling control the formation of complexes among proteins proximal to the TCR, the activation of kinase cascades, and the activation of transcription factors; however, the mode and extent of the influence of phosphorylation in coordinating the diverse phenomena associated with T cell activation are unclear. Therefore, we used the human Jurkat T cell leukemia cell line as a model system and performed large-scale quantitative phosphoproteomic analyses of TCR signaling. We identified 10,665 unique phosphorylation sites, of which 696 showed TCR-responsive changes. In addition, we analyzed broad trends in phosphorylation data sets to uncover underlying mechanisms associated with T cell activation. We found that, upon stimulation of the TCR, phosphorylation events extensively targeted protein modules involved in all of the salient phenomena associated with T cell activation: patterning of surface proteins, endocytosis of the TCR, formation of the F-actin cup, inside-out activation of integrins, polarization of microtubules, production of cytokines, and alternative splicing of messenger RNA. Further, case-by-case analysis of TCR-responsive phosphorylation sites on proteins belonging to relevant functional modules together with network analysis allowed us to deduce that serine-threonine (S-T) phosphorylation modulated protein-protein interactions (PPIs) in a system-wide fashion. We also provide experimental support for this inference by showing that phosphorylation of tubulin on six distinct serine residues abrogated PPIs during the assembly of microtubules. We propose that modulation of PPIs by stimulus-dependent changes in S-T phosphorylation state is a widespread phenomenon applicable to many other signaling systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
L-Milch-Dehydrogenase aus Kaninchenmuskel, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Glyceraldehyd-3-Phosphat-Dehydrogenase aus Kaninchenmuskel, lyophilized powder, ≥75 units/mg protein
Sigma-Aldrich
L-Milch-Dehydrogenase aus Kaninchenmuskel, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Milch-Dehydrogenase aus Rinderherz, 1000 units/mL
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type III, lyophilized powder, 350-600 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type II, ammonium sulfate suspension, 350-600 units/mg protein
Sigma-Aldrich
L-Milch-Dehydrogenase aus Rinderherz, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
Phosphoglucose-Isomerase aus Backhefe (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)
Sigma-Aldrich
Enolase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Aldolase aus Kaninchenmuskel, lyophilized powder, ≥8.0 units/mg protein
Sigma-Aldrich
3-Phosphoglycerin-Phosphokinase aus Backhefe (S. cerevisiae), ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
Pyruvat-Kinase aus Kaninchenmuskel, Type VII, buffered aqueous glycerol solution, 350-600 units/mg protein
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Schweineherz, ≥600 units/mg protein (biuret), ammonium sulfate suspension
Supelco
GAPDH, standard for protein electrophoresis
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Schweineherz, buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)
Sigma-Aldrich
Phosphoglucomutase aus Kaninchenmuskel, ammonium sulfate suspension, ≥100 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type X, lyophilized powder, ≥3,500 units/mg protein
Sigma-Aldrich
L-Milch-Dehydrogenase aus Rinderherz, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactat-Dehydrogenase, rekombinant aus E. coli, ≥90 U/mg
Sigma-Aldrich
Aldolase aus Kaninchenmuskel, ammonium sulfate suspension, 10-20 units/mg protein
Sigma-Aldrich
L-Milch-Dehydrogenase aus Schweineherz, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type III-S, ammonium sulfate suspension, ≥4,000 units/mg protein
Sigma-Aldrich
Phosphoglucose-Isomerase aus Kaninchenmuskel, Type XI, lyophilized powder, ≥200 units/mg protein
Sigma-Aldrich
Adenosine 5′-Triphosphatase from porcine cerebral cortex, lyophilized powder, ≥0.3 units/mg protein, pH 7.8
Sigma-Aldrich
Triosephosphate Isomerase from baker′s yeast (S. cerevisiae), Type I, ammonium sulfate suspension, ~10,000 units/mg protein
Sigma-Aldrich
Äpfelsäure-Dehydrogenase aus Rinderherz, ammonium sulfate suspension, 2000-4000 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
L-Milch-Dehydrogenase aus Rindermuskel, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Acetyl-CoA carboxylase 2 human, recombinant, expressed in Sf9 cells