Merck
  • RNA-binding protein FXR1 drives cMYC translation by recruiting eIF4F complex to the translation start site.
RNA-binding protein FXR1 drives cMYC translation by recruiting eIF4F complex to the translation start site.
Cell reports (2021-11-04)
Jasmine George, Yongsheng Li, Ishaque P Kadamberi, Deepak Parashar, Shirng-Wern Tsaih, Prachi Gupta, Anjali Geethadevi, Changliang Chen, Chandrima Ghosh, Yunguang Sun, Sonam Mittal, Ramani Ramchandran, Hallgeir Rui, Gabriel Lopez-Berestein, Cristian Rodriguez-Aguayo, Gustavo Leone, Janet S Rader, Anil K Sood, Madhusudan Dey, Sunila Pradeep, Pradeep Chaluvally-Raghavan
ZUSAMMENFASSUNG
Fragile X-related protein-1 (FXR1) gene is highly amplified in patients with ovarian cancer, and this amplification is associated with increased expression of both FXR1 mRNA and protein. FXR1 expression directly associates with the survival and proliferation of cancer cells. Surface sensing of translation (SUnSET) assay demonstrates that FXR1 enhances the overall translation in cancer cells. Reverse-phase protein array (RPPA) reveals that cMYC is the key target of FXR1. Mechanistically, FXR1 binds to the AU-rich elements (ARE) present within the 3' untranslated region (3'UTR) of cMYC and stabilizes its expression. In addition, the RGG domain in FXR1 interacts with eIF4A1 and eIF4E proteins. These two interactions of FXR1 result in the circularization of cMYC mRNA and facilitate the recruitment of eukaryotic translation initiation factors to the translation start site. In brief, we uncover a mechanism by which FXR1 promotes cMYC levels in cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Duolink® In Situ Detektionsreagenzien Rot
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Mitomycin C aus Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Puromycin-Antikörper, Klon 12D10, clone 12D10, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Magna RIP® RNA-Bindungsprotein Immunpräzipitations-Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cycloheximid, InSolution, 100 mg/mL in DMSO, Suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
GST protein, tag-free, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Puromycin, clone 12D10, Alexa Fluor 488 Conjugate Antibody, clone 12D10, 0.5 mg/mL, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-eIF4E in Kaninchen hergestellte Antikörper, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.