Merck
  • Pharmacological inhibition of nicotinamide phosphoribosyltransferase/visfatin enzymatic activity identifies a new inflammatory pathway linked to NAD.
Pharmacological inhibition of nicotinamide phosphoribosyltransferase/visfatin enzymatic activity identifies a new inflammatory pathway linked to NAD.
PloS one (2008-05-22)
Nathalie Busso, Mahir Karababa, Massimo Nobile, Aline Rolaz, Frédéric Van Gool, Mara Galli, Oberdan Leo, Alexander So, Thibaut De Smedt
ZUSAMMENFASSUNG
Nicotinamide phosphoribosyltransferase (NAMPT), also known as visfatin, is the rate-limiting enzyme in the salvage pathway of NAD biosynthesis from nicotinamide. Since its expression is upregulated during inflammation, NAMPT represents a novel clinical biomarker in acute lung injury, rheumatoid arthritis, and Crohn's disease. However, its role in disease progression remains unknown. We report here that NAMPT is a key player in inflammatory arthritis. Increased expression of NAMPT was confirmed in mice with collagen-induced arthritis, both in serum and in the arthritic paw. Importantly, a specific competitive inhibitor of NAMPT effectively reduced arthritis severity with comparable activity to etanercept, and decreased pro-inflammatory cytokine secretion in affected joints. Moreover, NAMPT inhibition reduced intracellular NAD concentration in inflammatory cells and circulating TNFalpha levels during endotoxemia in mice. In vitro pharmacological inhibition of NAMPT reduced the intracellular concentration of NAD and pro-inflammatory cytokine secretion by inflammatory cells. Thus, NAMPT links NAD metabolism to inflammatory cytokine secretion by leukocytes, and its inhibition might therefore have therapeutic efficacy in immune-mediated inflammatory disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3,3′-Diaminobenzidin, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by gel-filtration chromatography
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by trichloroacetic acid extraction
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3,3′-Diaminobenzidin, 97% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, FITC conjugate
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, Detoxified
Preise und Verfügbarkeit sind derzeit nicht verfügbar.