Merck
  • Bifidobacterium strains suppress in vitro the pro-inflammatory milieu triggered by the large intestinal microbiota of coeliac patients.
Bifidobacterium strains suppress in vitro the pro-inflammatory milieu triggered by the large intestinal microbiota of coeliac patients.
Journal of inflammation (London, England) (2008-11-05)
Marcela Medina, Giada De Palma, Carmen Ribes-Koninckx, Miguel Calabuig, Yolanda Sanz
ZUSAMMENFASSUNG
Coeliac disease (CD) is an enteropathy characterized by an aberrant immune response to cereal-gluten proteins. Although gluten peptides and microorganisms activate similar pro-inflammatory pathways, the role the intestinal microbiota may play in this disorder is unknown. The purpose of this study was to assess whether the faecal microbiota of coeliac patients could contribute to the pro-inflammatory milieu characteristic of CD and the possible benefits of bifidobacteria. The effect of faeces of 26 CD patients with active disease (mean age 5.5 years, range 2.1-12.0 years), 18 symptom-free coeliac disease (SFCD) patients (mean age 5.5 years, range 1.0-12.3 years) on a gluten-free diet for 1-2 years; and 20 healthy children (mean age 5.3 years, range 1.8-10.8 years) on induction of cytokine production and surface antigen expression in peripheral blood mononuclear cells (PBMCs) were determined. The possible regulatory roles of Bifidobacterium longum ES1 and B. bifidum ES2 co-incubated with faecal samples were also assessed in vitro. Faeces of both active CD and SFCD patients, representing an imbalanced microbiota, significantly increased TNF-alpha production and CD86 expression in PBMCs, while decreased IL-10 cytokine production and CD4 expression compared with control samples. Active CD-patient samples also induced significantly higher IFN-gamma production compared with controls. However, Bifidobacterium strains suppressed the pro-inflammatory cytokine pattern induced by the large intestinal content of CD patients and increased IL-10 production. Cytokine effects induced by faecal microbiota seemed to be mediated by the NFkappaB pathway. The intestinal microbiota of CD patients could contribute to the Th1 pro-inflammatory milieu characteristic of the disease, while B. longum ES1 and B. bifidum ES2 could reverse these deleterious effects. These findings hold future perspectives of interest in CD therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, Ready Made solution, 1 mg/mL
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by trichloroacetic acid extraction
SAFC
L-Cystein
Sigma-Aldrich
D-Cystein, ≥99% (RT)
Sigma-Aldrich
L-Cystein, ≥97%, FG
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, FITC conjugate
Supelco
L-Cystein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystein, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Lactacystin, ≥90% (HPLC)
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, Detoxified