Merck
  • The structure of human microplasmin in complex with textilinin-1, an aprotinin-like inhibitor from the Australian brown snake.
The structure of human microplasmin in complex with textilinin-1, an aprotinin-like inhibitor from the Australian brown snake.
PloS one (2013-01-22)
Emma-Karin I Millers, Lambro A Johnson, Geoff W Birrell, Paul P Masci, Martin F Lavin, John de Jersey, Luke W Guddat
ZUSAMMENFASSUNG
Textilinin-1 is a Kunitz-type serine protease inhibitor from Australian brown snake venom. Its ability to potently and specifically inhibit human plasmin (K(i) = 0.44 nM) makes it a potential therapeutic drug as a systemic anti-bleeding agent. The crystal structures of the human microplasmin-textilinin-1 and the trypsin-textilinin-1 complexes have been determined to 2.78 Å and 1.64 Å resolution respectively, and show that textilinin-1 binds to trypsin in a canonical mode but to microplasmin in an atypical mode with the catalytic histidine of microplasmin rotated out of the active site. The space vacated by the histidine side-chain in this complex is partially occupied by a water molecule. In the structure of microplasminogen the χ(1) dihedral angle of the side-chain of the catalytic histidine is rotated by 67° from its "active" position in the catalytic triad, as exemplified by its location when microplasmin is bound to streptokinase. However, when textilinin-1 binds to microplasmin the χ(1) dihedral angle of this amino acid residue changes by -157° (i.e. in the opposite rotation direction compared to microplasminogen). The unusual mode of interaction between textilinin-1 and plasmin explains textilinin-1's selectivity for human plasmin over plasma kallikrein. This difference can be exploited in future drug design efforts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Aprotinin aus Rinderlunge, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Aprotinin aus Rinderlunge, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, Type II-S, 1,000-2,000 units/mg dry solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, powder, ≥7,500 BAEE units/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Aprotinin aus Rinderlunge, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Aprotinin aus Rinderlunge, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Trypsin aus Rinderpankreas, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Plasmin aus Humanplasma, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Aprotinin aus Rinderlunge
Sigma-Aldrich
Trypsin aus Rinderpankreas, essentially salt-free, lyophilized powder, ≥9,000 BAEE units/mg protein, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin aus Rinderpankreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Trypsin aus Schweinepankreas, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Aprotinin aus Rinderlunge, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg
Sigma-Aldrich
Trypsin aus Rinderpankreas, Type XI, lyophilized powder, ≥6,000 BAEE units/mg protein
Sigma-Aldrich
Aprotinin aus Rinderlunge, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
TrypZean® Rind, recombinant, expressed in corn, lyophilized powder, ≥3,350 units/mg solid (USP)
Sigma-Aldrich
Trypsin aus Schweinepankreas, tablet, 1 mg tablet
Sigma-Aldrich
Trypsin aus Schweinepankreas, ~1500 U/mg
Sigma-Aldrich
Trypsin Acetylated from bovine pancreas, Type V-S, ≥8,500 BAEE units/mg protein (biuret)
Sigma-Aldrich
Trypsin aus meschlichem Pankreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Sigma-Aldrich
Kallikrein from human plasma, buffered aqueous solution, ≥5 units/mg protein
Sigma-Aldrich
Trypsin aus Rinderpankreas, ≥2,500 USP units/mg solid, meets USP testing specifications