Merck
  • The Arabidopsis thaliana ortholog of a purported maize cholinesterase gene encodes a GDSL-lipase.
The Arabidopsis thaliana ortholog of a purported maize cholinesterase gene encodes a GDSL-lipase.
Plant molecular biology (2013-02-23)
Mrinalini Muralidharan, Kristina Buss, Katherine E Larrimore, Nicholas A Segerson, Latha Kannan, Tsafrir S Mor
ZUSAMMENFASSUNG
Acetylcholinesterase is an enzyme that is intimately associated with regulation of synaptic transmission in the cholinergic nervous system and in neuromuscular junctions of animals. However the presence of cholinesterase activity has been described also in non-metazoan organisms such as slime molds, fungi and plants. More recently, a gene purportedly encoding for acetylcholinesterase was cloned from maize. We have cloned the Arabidopsis thaliana homolog of the Zea mays gene, At3g26430, and studied its biochemical properties. Our results indicate that the protein encoded by the gene exhibited lipase activity with preference to long chain substrates but did not hydrolyze choline esters. The At3g26430 protein belongs to the SGNH clan of serine hydrolases, and more specifically to the GDS(L) lipase family.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipase aus Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase aus Schweinepankreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase-Acrylharz aus Candida antarctica, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase aus Schweinepankreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Lipase aus Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase aus Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase aus Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase, immobilisiert aus Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥900 units/mg protein
Sigma-Aldrich
Lipase aus Rhizomucor miehei, ≥20,000 U/g
Sigma-Aldrich
Lipase aus Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Butyrylcholinesterase aus Humanserum, vial of ≥4 units
Sigma-Aldrich
Lipase aus Weizenkeimen, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase aus Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase aus Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase aus Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase aus Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Butyrylcholinesterase aus Pferdeserum, lyophilized powder, ≥500 units/mg protein
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase aus Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase aus Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)