Merck
  • Novel organic solvent-tolerant esterase isolated by metagenomics: insights into the lipase/esterase classification.
Novel organic solvent-tolerant esterase isolated by metagenomics: insights into the lipase/esterase classification.
Revista Argentina de microbiologia (2013-04-09)
Renaud Berlemont, Olivier Spee, Maud Delsaute, Yannick Lara, Jörg Schuldes, Carola Simon, Pablo Power, Rolf Daniel, Moreno Galleni
ZUSAMMENFASSUNG
in order to isolate novel organic solvent-tolerant (OST) lipases, a metagenomic library was built using DNA derived from a temperate forest soil sample. A two-step activity-based screening allowed the isolation of a lipolytic clone active in the presence of organic solvents. Sequencing of the plasmid pRBest recovered from the positive clone revealed the presence of a putative lipase/esterase encoding gene. The deduced amino acid sequence (RBest1) contains the conserved lipolytic enzyme signature and is related to the previously described OST lipase from Lysinibacillus sphaericus 205y, which is the sole studied prokaryotic enzyme belonging to the 4.4 α/β hydrolase subgroup (abH04.04). Both in vivo and in vitro studies of the substrate specificity of RBest1, using triacylglycerols or nitrophenyl-esters, respectively, revealed that the enzyme is highly specific for butyrate (C4) compounds, behaving as an esterase rather than a lipase. The RBest1 esterase was purified and biochemically characterized. The optimal esterase activity was observed at pH 6.5 and at temperatures ranging from 38 to 45 °C. Enzymatic activity, determined by hydrolysis of p-nitrophenyl esters, was found to be affected by the presence of different miscible and non-miscible organic solvents, and salts. Noteworthy, RBest1 remains significantly active at high ionic strength. These findings suggest that RBest1 possesses the ability of OST enzymes to molecular adaptation in the presence of organic compounds and resistance of halophilic proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipase aus Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase aus Schweinepankreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase-Acrylharz aus Candida antarctica, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase aus Schweinepankreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase aus Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase aus Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase aus Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase, immobilisiert aus Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase aus Rhizomucor miehei, ≥20,000 U/g
Sigma-Aldrich
Lipase aus Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase aus Weizenkeimen, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase aus Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase aus Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase aus Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase aus Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase aus Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase aus Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)