Merck
  • Relationship between the expression of CES2, UGT1A1, and GUSB in colorectal cancer tissues and aberrant methylation.
Relationship between the expression of CES2, UGT1A1, and GUSB in colorectal cancer tissues and aberrant methylation.
Neoplasma (2013-11-08)
F W Xie, Y Peng, Xiong Chen, Xi Chen, J Li, W Wang, Z Yu, X Ouyang
ZUSAMMENFASSUNG
Irinotecan (CPT-11) is considered an important drug in the treatment of colorectal cancer, but its continuous administration reduces its sensitivity and influences the curative effect. The metabolism of CPT-11 is mainly controlled by carboxy-lesterase (CES), UDP-glucuronosyltransferase 1A (UGT1A), and β-glucuronidase (GUSB). Studies to date have shown that methylation acts as an important mechanism for gene expression to suppress the metabolic enzymes of many chemotherapeutics. This study, which selected 99 colorectal cancer patients, 23 of whom had paracancerous tissues and eight of whom had large intestine adenomas, aimed to investigate the correlation between the protein expression of the CPT-11 metabolic enzyme genes CES2, UGT1A1, and GUSB and various clinical pathological parameters of colorectal cancer tissues, as well as the relationship between methylation regulation and the gene expression of CES2, UGT1A1, and GUSB. We used immunohistochemistry staining, methylation-specific PCR, and clinical status to reveal the possible regulatory targets of chemotherapeutic resistance in colorectal cancer and to provide new ideas and countermeasures to reverse anti-cancer drug resistance and chemosensitization. The results showed that the expression of CES2, UGTA1A1, and GUSB varies in colorectal pathology tissues and that the expression of CES2 is somewhat related to tumor staging. This relationship is likely caused by the gene regulation of UGT1A1 and GUSB, and other regulation mechanisms may also be involved. The methylation of the CES2 gene is irrelevant to the morbidity associated with colorectal cancer. The GUSB gene showed no significant differences in methylation, and the hemi-methylation was also positive, the regulating ability of which needs to be verified. The potential role of these genes in the colorectal cancer progression, which may be directly related to the methylation regulation of UGT1A1, requires further research. The promoter of the UGT1A1 gene in colorectal cancer cells is methylated, which is an important mechanism of UGT1A1 gene silencing and can be regarded as the target point of research for CPT-11 drug resistance and control mechanisms for the reversal of drug resistance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
β-Glucuronidase aus Napfschnecke (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Rinderleber, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
Esterase aus Schweineleber, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
Esterase aus Schweineleber, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
Esterase aus Kaninchenleber, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
ββ-Glucuronidase aus E. coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
β-Glucuronidase aus Rinderleber, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase aus Napfschnecke (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
β-Glucuronidase aus Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
Esteraseisoenzym-1 aus Schweineleber, rekombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4