Merck
  • Genetic variants in GCKR and PNPLA3 confer susceptibility to nonalcoholic fatty liver disease in obese individuals.
Genetic variants in GCKR and PNPLA3 confer susceptibility to nonalcoholic fatty liver disease in obese individuals.
The American journal of clinical nutrition (2014-01-31)
Yu-Cheng Lin, Pi-Feng Chang, Mei-Hwei Chang, Yen-Hsuan Ni
ZUSAMMENFASSUNG
A genome-wide association study identified variants in or near patatin-like phospholipase domain-containing-3 (PNPLA3), neurocan (NCAN), lysophospholipase-like 1 (LYPLAL1), glucokinase regulatory protein (GCKR), and protein phosphatase 1 regulatory subunit 3b (PPP1R3B) that were strongly associated with nonalcoholic fatty liver disease (NAFLD) in adults of European ancestry. We examined these genetic variants in obese children and tested whether their effects on NAFLD are significant in the Taiwanese Han Chinese population. We genotyped PNPLA3 rs738409, NCAN rs2228603, LYPLAL1 rs12137855, GCKR rs780094, and PPP1R3B rs4240624 in 797 obese children aged 7-18 y. NAFLD was identified by liver ultrasonography. We analyzed the effect of these genetic variants on NAFLD. NAFLD was identified in 24% of the recruited obese children. We found significant associations with NAFLD at variants in PNPLA3 and GCKR but not in NCAN, LYPLAL1, and PPP1R3B. Multiple logistic regression analysis showed that, after control for the effects of age- and sex-adjusted body mass index, waist-to-hip ratio, sex, and PNPLA3 rs738409 polymorphism, the variant GCKR rs780094 TT genotype independently increased the OR of NAFLD by 1.997 (95% CI: 1.196, 3.335; P = 0.008) compared with the CC genotype. Subjects with the variant GCKR rs780094 TT genotype had a higher mean serum alanine aminotransferase concentration than did those with the CC genotype (30.8 ± 34.7 compared with 22.2 ± 18.6 IU/L; P = 0.01). By studying the genetic variants of obese Taiwanese children, we confirmed that the genetic variants in GCKR rs780094 and PNPLA3 rs738409, but not in NCAN rs2228603, LYPLAL1 rs12137855, and PPP1R3B rs4240624, are associated with an increased risk of NAFLD. GCKR and PNPLA3 variants are the common genetic factors that may confer susceptibility to NAFLD in obese individuals across multiple ethnic groups.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipase aus Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase aus Schweinepankreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase-Acrylharz aus Candida antarctica, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase aus Schweinepankreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase aus Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase aus Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase aus Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase, immobilisiert aus Candida antarctica, beads, slightly brown, >2 U/mg
Sigma-Aldrich
Lipase aus Rhizomucor miehei, ≥20,000 U/g
Sigma-Aldrich
Lipase aus Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
Lipase aus Weizenkeimen, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase aus Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase aus Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase aus Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase aus Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase aus Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase aus Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, rekombinant aus Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase aus Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase aus Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)