Merck
  • Early specification of CD8+ T lymphocyte fates during adaptive immunity revealed by single-cell gene-expression analyses.
Early specification of CD8+ T lymphocyte fates during adaptive immunity revealed by single-cell gene-expression analyses.
Nature immunology (2014-03-04)
Janilyn Arsenio, Boyko Kakaradov, Patrick J Metz, Stephanie H Kim, Gene W Yeo, John T Chang
ZUSAMMENFASSUNG
T lymphocytes responding to microbial infection give rise to effector cells that mediate acute host defense and memory cells that provide long-lived immunity, but the fundamental question of when and how these cells arise remains unresolved. Here we combined single-cell gene-expression analyses with 'machine-learning' approaches to trace the transcriptional 'roadmap' of individual CD8(+) T lymphocytes throughout the course of an immune response in vivo. Gene-expression signatures predictive of eventual fates could be discerned as early as the first T lymphocyte division and may have been influenced by asymmetric partitioning of the receptor for interleukin 2 (IL-2Rα) during mitosis. Our findings emphasize the importance of single-cell analyses in understanding fate determination and provide new insights into the specification of divergent lymphocyte fates early during an immune response to microbial infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Albumin aus Hühnereiweiss, powder, 62-88% (agarose gel electrophoresis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥90% (agarose gel electrophoresis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Albumin aus Hühnereiweiss, For use as a marker in SDS-PAGE
Preise und Verfügbarkeit sind derzeit nicht verfügbar.