Merck
  • Role of rhBMP-2 and rhBMP-7 in the metabolism and differentiation of osteoblast-like cells cultured on chemically modified titanium surfaces.
Role of rhBMP-2 and rhBMP-7 in the metabolism and differentiation of osteoblast-like cells cultured on chemically modified titanium surfaces.
The Journal of oral implantology (2014-12-17)
Fabiano Ribeiro Cirano, Adriane Yaeko Togashi, Márcia Martins Marques, Francisco Emílio Pustiglioni, Luiz Antônio Pugliesi Alves Lima
ZUSAMMENFASSUNG
This study analyzed the role of recombinant human bone morphogenetic protein 2 (rhBMP-2) and recombinant human bone morphogenetic protein 7 (rhBMP-7) in the adhesion and differentiation of rat osteoblast-like (osteo-1) cells cultured on chemically modified titanium surfaces. Osteo-1 cells were cultured on chemically modified (modified sandblasted and acid-etched) titanium surfaces in 3 different types of medium: control, medium supplemented with 20 ng/mL rhBMP-2, and medium supplemented with 20 ng/mL rhBMP-7. The following parameters were evaluated: cell adhesion after 24 hours; total protein content; collagen content and alkaline phosphatase (AP) activity after 7, 14, and 21 days; and calcified nodule formation after 21 days. The addition of rhBMP-2 or rhBMP-7 did not influence cell adhesion (P = .1175). Cell differentiation was influenced by rhBMP-2, as demonstrated by a significant increase in collagen content after 7 days of culture (P < .0001) and a significant decrease in AP activity after 21 days (P < .0001). The addition of rhBMP-7 only influenced AP activity, and a significant increase was observed after 21 days (P < .0001). Within the limitations of the study, we conclude that the presence of rhBMP-2 or rhBMP-7 did not influence cell adhesion to chemically modified titanium surfaces but provided an additional stimulus during the differentiation of rat osteo-1 cells cultured on this type of surface.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen, Typ I -Lösung aus Rattenschwanz, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Kollagen aus Rattenschwanz, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Achillessehne vom Rind, powder, suitable for substrate for collagenase
Sigma-Aldrich
Kollagen aus chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus Kalbshaut, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Kollagen Typ IV aus human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Titan, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Kollagen aus bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Titan, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titan, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Titan, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
Kollagen aus bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Kollagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Kollagen aus menschlicher Placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Kollagen aus Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Titan, mesh, 100x100mm, nominal aperture 0.19mm, wire diameter 0.23mm, 60x60 wires/inch, open area 20%, twill weave