Merck
Factors affecting ultrasonic release from eLiposomes.
Journal of pharmaceutical sciences (2015-02-03)
James R Lattin, William G Pitt
ZUSAMMENFASSUNG
Liposomes containing emulsion droplets (eLiposomes) were studied as ultrasound-responsive liposomal drug carriers. This paper presents the effects of temperature, eLiposome size, and ultrasound parameters on the ultrasonically actuated release of calcein, to test hypotheses concerning the physics of acoustic droplet vaporization with regard to vapor pressure and Laplace pressure. Small (200 nm) eLiposomes containing 100-nm emulsion droplets were formed and compared with large (800 nm) eLiposomes containing 100- or 450-nm droplets. Calcein release was quantified by spectroscopic methods. Various experiments examined the influence of perfluorocarbon (PFC) droplet size, vesicle size, temperature, PFC composition and vapor pressure, insonation time, and insonation frequency. Results showed that eLiposome samples released significantly more calcein than their conventional liposome counterparts. Surprisingly, temperature (which directly controls vapor pressure) did not have a strong effect on ultrasound-induced calcein release. In general, calcein release decreased with decreasing droplet size, as hypothesized based on Laplace pressure. Release decreased with increased ultrasound frequency if the pressure amplitude and exposure time were maintained constant, indicating that the gas-phase nucleation rate may have an important role in rupture of eLiposomes. Interestingly, when ultrasound of the same mechanical index was applied at two frequencies, the amount of release correlated strongly with the mechanical index.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tetradecafluorhexan, 99%
Supelco
Chloroform, analytical standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetradecafluorhexan, A mixture of perfluorinated hexanes, 95%