Merck
Increased beta 2 defensin in recurrent aphthous ulcer.
Oral diseases (2014-05-24)
A Al-Samadi, A Salem, M Ainola, J Hietanen, R Häyrinen-Immonen, Y T Konttinen
ZUSAMMENFASSUNG
It was hypothesized that beta 2 defensin (BD-2) is increased in RAU lesions compared with healthy controls to promote anti-microbial host defence. RAU and control mucosa samples were subjected to quantitative real-time PCR and immunostained for BD-2, CD68, mast cell tryptase and 4-hydroxynonenal (4HNE). The effect of tumour necrosis factor-α (TNF-α) ± interleukin-17C (IL-17C), without and with vitamin K3, was studied on BD-2 expression in epithelial SCC-25 cells. Although BD-2 mRNA did not differ between healthy and RAU mucosa, BD-2 stained strongly in acute-phase RAU epithelium (P = 0.001). In controls, subepithelial BD-2(+) cells were mast cells and macrophages, whereas in RAU, most infiltrating leucocytes were BD-2(+) (P = 0.004). In cell culture, BD-2 was increased 124-fold by TNF-α (P < 0.0001) and 208-fold synergistically together with IL-17C (P < 0.0001). 4HNE staining of RAU epithelium was not significantly increased, and vitamin K3-induced reactive oxygen species (ROS) did not affect BD-2. Anti-microbial BD-2 was not affected by oxidative stress but was highly increased in the epithelial and immigrant cells in the acute-phase RAU lesions, probably in part synergistically by TNF-α and epithelial IL-17C, which are known to be induced by activation of danger-signal receptors by pathogen- and/or damage-associated molecular patterns.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Menadion, crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Menadion, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,2,4,5-Tetramethylbenzen, 95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Menadion (K3), analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
1,2,4,5-Tetramethyl-benzol, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Menadion, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Menadion, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Menadion, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.