Merck
  • Safety evaluation of a triazine compound nitromezuril by assessing bacterial reverse mutation, sperm abnormalities, micronucleus and chromosomal aberration.
Safety evaluation of a triazine compound nitromezuril by assessing bacterial reverse mutation, sperm abnormalities, micronucleus and chromosomal aberration.
Regulatory toxicology and pharmacology : RTP (2015-02-04)
Chenzhong Fei, Jie Zhang, Yang Lin, Xiaoyang Wang, Keyu Zhang, Lifang Zhang, Wenli Zheng, Mi Wang, Tao Li, Sui Xiao, Feiqun Xue, Chunmei Wang
ZUSAMMENFASSUNG
Nitromezuril (NZL) is a novel triazine compound that exhibits remarkable anticoccidial activity. However, mutagenicity and genotoxicity of NZL have not been evaluated to date. This study evaluated the potential risks of NZL by testing for bacterial reverse mutation (Ames), mouse sperm abnormality (SA), bone marrow micronucleus (MN) and chromosomal aberration (CA). Mice were orally administered with NZL at 385, 192 and 96 mg/kg, corresponding to 0.5 ×, 0.25 × and 0.125 × the LD50 of NZL, respectively. No significant increases in SA and CA were found in mice treated with NZL for 5d and 3d, respectively (P>0.05). NZL at 96-385 mg/kg did not have significant influence on micronucleated polychromatic erythrocyte counts (P>0.05). These results suggest that NZL is not genotoxic. However, Ames test results were positive both with and without the S9 system for Salmonella typhimurium TA98 and TA100, suggesting that NZL may be mutagenic. The mutagenic effects of NZL were different in in vitro and in vivo assays. Further studies should be conducted to confirm the safety of using and developing NZL as a novel anticoccidial drug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Hydrat
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
2-Aminoanthracen, 96%
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 5 mg (per vial)
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
2-Aminofluoren, 98%
Cyclophosphamid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fenaminosulf, ≥98%
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
β-Nicotinamidadenin-Dinucleotidphosphat Natriumsalz, pkg of 10 mg (per vial)
Sigma-Aldrich
N6-Cyclopentyladenosine, solid
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Supelco
Fenaminosulf, PESTANAL®, analytical standard