Merck
Ebp1 activates podoplanin expression and contributes to oral tumorigenesis.
Oncogene (2013-08-27)
Y Mei, P Zhang, H Zuo, D Clark, R Xia, J Li, Z Liu, L Mao
ZUSAMMENFASSUNG
Podoplanin is highly expressed in human cancers. However, mechanisms regulating podoplanin expression remain elusive. Here we show that podoplanin promotes tumorigenesis of oral squamous cell carcinoma (OSCC) and precancerous cells both in vitro and in vivo, and the ErbB3-binding protein-1 (Ebp1) can be activated in oral tumorigenesis and can serve as a transcriptional activator to drive podoplanin expression in the malignant progression. Most of the OSCC cell lines have no detectable podoplanin protein in low-density cultures. However, the protein becomes detectable in high-density cultures and is required for in-vivo tumor formation of OSCC and oral premalignancies. In a high-density culture condition, podoplanin expression can be triggered at both mRNA and protein levels. In this condition, we showed that Ebp1 is upregulated, translocated from the cytoplasm to the nucleus and binds to the podoplanin promoter to result in a dramatic increase of podoplanin mRNA and protein. Ebp1 downregulation significantly reduced podoplanin expression levels in OSCC cells with a decreased anchorage-dependent growth, invasion and wound healing. Conversely, Ebp1 overexpression enhanced these malignant features through podoplanin upregulation both in vitro and in vivo. In 81 patients with oral premalignant lesions, we found that Ebp1 expression is strongly related to OSCC development. We conclude that Ebp1 has a key role in the upregulation of podoplanin and may contribute to oral tumorigenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Jodacetamid
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis