Merck
  • Development of a new sorptive extraction method based on simultaneous direct and headspace sampling modes for the screening of polycyclic aromatic hydrocarbons in water samples.
Development of a new sorptive extraction method based on simultaneous direct and headspace sampling modes for the screening of polycyclic aromatic hydrocarbons in water samples.
Talanta (2014-12-06)
Sara Triñanes, Ma Teresa Pena, Ma Carmen Casais, Ma Carmen Mejuto
ZUSAMMENFASSUNG
A new straightforward and inexpensive sample screening method for both EPA and EU priority polycyclic aromatic hydrocarbons (PAHs) in water has been developed. The method is based on combined direct immersion and headspace (DIHS) sorptive extraction, using low-cost disposable material, coupled to ultraperformance liquid chromatography with fluorescence and UV detection (UPLC-FD-UV). Extraction parameters, such as the sampling mode, extraction time and ionic strength were investigated in detail and optimized. Under optimized conditions, water samples (16 mL) were concentrated in silicone disks by headspace (HS) and direct immersion (DI) modes simultaneously, at room temperature for 9h for the majority of the 24 studied compounds. Ultrasound-assisted desorption of extracted analytes in acetonitrile was carried out also at room temperature. The optimized chromatographic method provided a good linearity (R≥0.9991) and a broad linear range for all studied PAHs. The proposed analytical procedure exhibited a good precision level with relative standard deviations below 15% for all analytes. Quantification limits between 0.7 and 2.3 µg L(-1) and 0.16 and 3.90 ng L(-1) were obtained for compounds analyzed by UV (acenaphtylene, cyclopenta[c,d]pyrene and benzo[j]fluoranthene) and fluorescence, respectively. Finally, the proposed method was applied to the determination of PAHs in different real tap, river and wastewater samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Methanol, Absolute - Acetone free