Merck
  • Polylactide-based paclitaxel-loaded nanoparticles fabricated by dispersion polymerization: characterization, evaluation in cancer cell lines, and preliminary biodistribution studies.
Polylactide-based paclitaxel-loaded nanoparticles fabricated by dispersion polymerization: characterization, evaluation in cancer cell lines, and preliminary biodistribution studies.
Journal of pharmaceutical sciences (2014-06-26)
Simeon K Adesina, Alesia Holly, Gabriela Kramer-Marek, Jacek Capala, Emmanuel O Akala
ZUSAMMENFASSUNG
The macromonomer method was used to prepare cross-linked, paclitaxel-loaded polylactide (PLA)-polyethylene glycol (stealth) nanoparticles using free-radical dispersion polymerization. The method can facilitate the attachment of other molecules to the nanoparticle surface to make it multifunctional. Proton nuclear magnetic resonance and Fourier transform infrared spectra confirm the synthesis of PLA macromonomer and cross-linking agent. The formation of stealth nanoparticles was confirmed by scanning and transmission electron microscopy. The drug release isotherm of paclitaxel-loaded nanoparticles shows that the encapsulated drug is released over 7 days. In vitro cytotoxicity assay in selected breast and ovarian cancer cell lines reveal that the blank nanoparticle is biocompatible compared with medium-only treated controls. In addition, the paclitaxel-loaded nanoparticles exhibit similar cytotoxicity compared with paclitaxel in solution. Confocal microscopy reveals that the nanoparticles are internalized by MCF-7 breast cancer cells within 1 h. Preliminary biodistribution studies also show nanoparticle accumulation in tumor xenograft model. The nanoparticles are suitable for the controlled delivery of bioactive agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Hydroxylamin -hydrochlorid, ReagentPlus®, 99%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Hydroxylamin -hydrochlorid, ACS reagent, 98.0%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
2-Hydroxyethylmethacrylat, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
2-Hydroxyethylmethacrylat, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Hydroxylamin -hydrochlorid, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methacryloylchlorid, 97%, contains monomethyl ether hydroquinone as stabilizer
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Hydroxylamin -hydrochlorid, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Toluol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridin, ≥99%
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material