Merck
Sensitive and selective cocaine electrochemical detection using disposable sensors.
Analytica chimica acta (2014-06-15)
Laura Asturias-Arribas, M Asunción Alonso-Lomillo, Olga Domínguez-Renedo, M Julia Arcos-Martínez
ZUSAMMENFASSUNG
This paper describes the voltammetric determination of cocaine in presence of three different interferences that could be found in street samples using disposable sensors. The electrochemical analysis of this alkaloid can be affected by the presence of codeine, paracetamol or caffeine, whose oxidation peaks may overlap and lead to false positives. This work describes two different solutions to this problem. On one hand, the modification of disposable carbon sensors with carbon nanotubes allows the voltammetric quantification of cocaine by using ordinary least squares regressions in the concentration range from 10 to 155 μmol L(-1), with a reproducibility of 5.6% (RSD, n = 7. On the other hand, partial least squares regressions are used for the resolution of the overlapped voltammetric signals when using screen-printed carbon electrodes without any modification. Both procedures have been successfully applied to the evaluation of the purity of cocaine street samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Kaliumnitrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Supelco
4-Acetamidophenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Kaliumnitrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumnitrat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Aceton, analytical standard