Merck
  • Highly sensitive quantification of pyrethroid insecticide etofenprox in vegetables with high-performance liquid chromatography and fluorescence detection.
Highly sensitive quantification of pyrethroid insecticide etofenprox in vegetables with high-performance liquid chromatography and fluorescence detection.
Journal of chromatography. A (2015-02-11)
Eiki Watanabe, Koji Baba
ZUSAMMENFASSUNG
This paper describes a highly sensitive analytical method using high-performance liquid chromatography and fluorescence detection (HPLC-FLD) capable of quantifying trace amounts of synthetic pyrethroid insecticide etofenprox residue in six vegetable samples: bell pepper, cucumber, eggplant, Japanese mustard spinach, spinach, and tomato. After extraction with acetonitrile, the crude sample extract was cleaned up with a solid-phase extraction cartridge. The matrix interference derived from the tested vegetable samples was evaluated. Quantification was conducted using external calibrators prepared in pure acetonitrile. The limits of quantification for etofenprox in each sample were 1.87-3.87 ng/g. Recoveries obtained by application of the proposed analytical method of vegetable samples spiked at the considerably low levels (5-100 ng/g) were 85-111% with relative standard deviations of less than 12%. The proposed method using the HPLC-FLD was applied for trace analysis of the insecticide residue in vegetable samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumsulfat, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Magnesiumsulfat, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%