Merck
  • Simultaneous quantification of tobacco alkaloids and major phase I metabolites by LC-MS/MS in human tissue.
Simultaneous quantification of tobacco alkaloids and major phase I metabolites by LC-MS/MS in human tissue.
International journal of legal medicine (2014-10-12)
Lisa Fischer, Felix Mikus, Ricarda Jantos, Gisela Skopp
ZUSAMMENFASSUNG
Insurance agencies might request laboratories to differentiate whether a deceased has been a smoker or not to decide about refunding of his nonsmoker rate. In this context, the question on a solid proof of tobacco alkaloids and major metabolites in tissues came up. Currently, an appropriate assay is still lacking to analyze tissue distribution in smokers or nonsmokers. Nicotine (NIC), nornicotine (NNIC), anatabine (ATB), anabasine (ABS), and myosmine (MYO) are naturally occurring alkaloids of the tobacco plant; most important phase I metabolites of NIC are cotinine (COT), norcotinine (NCOT), trans-3'-hydroxycotinine (HCOT), nicotine-N'-oxide (NNO), and cotinine-N-oxide (CNO). An analytical assay for their determination was developed and applied to five randomly selected autopsy cases. Homogenates using 500 mg aliquots of tissue samples were analyzed by liquid chromatography/tandem mass spectrometry following solid phase extraction. The method was validated according to current international guidelines. NIC, COT, NCOT, ABS, ATB, and HCOT could be detected in all tissues under investigation. Highest NIC concentrations were observed in the lungs, whereas highest COT concentrations have been found in the liver. MYO was not detectable in any of the tissues under investigation. The assay is able to adequately separate isobaric analyte pairs such as NIC/ABS/NCOT and HCOT/CNO thus being suitable for the determination of tobacco alkaloids and their phase I metabolites from tissue. More autopsy cases as well as corresponding body fluids and hair samples will be investigated to differentiate smokers from nonsmokers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications