Merck
  • Natural compounds boldine and menthol are antagonists of human 5-HT3 receptors: implications for treating gastrointestinal disorders.
Natural compounds boldine and menthol are antagonists of human 5-HT3 receptors: implications for treating gastrointestinal disorders.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2014-04-09)
J Walstab, C Wohlfarth, R Hovius, S Schmitteckert, R Röth, F Lasitschka, M Wink, H Bönisch, B Niesler
ZUSAMMENFASSUNG
Impaired 5-HT3 receptor function is likely involved in the pathogenesis of functional gastrointestinal disorders (FGID) and 5-HT3 receptor antagonists are effective treatments for chemotherapy-induced nausea and vomiting (CINV) and irritable bowel syndrome (IBS). The monoterpene alcohol menthol and the aporphine alkaloid boldine combat symptoms of gastrointestinal diseases; both interact with other members of the Cys-loop ligand-gated ion channel family and may therefore also act on 5-HT3 receptors. The impact of boldine and menthol on human recombinant homomeric 5-HT3 A- and heteromeric 5-HT3 AB receptors in HEK293 cells was determined by radioligand binding, a luminescence-based Ca(2+) assay, and a membrane potential assay. 5-HT3 protein and mRNA expression was assessed in human colon tissue. Boldine and menthol inhibited the 5-HT-induced activation of 5-HT3 receptors in the low and middle micromolar range, respectively. Boldine was a competitive antagonist of both receptors being 6.5- to 10-fold more potent at 5-HT3 A- vs 5-HT3 AB receptors. Menthol non-competitively and stereoselectively inhibited both receptors: In contrast to (+)-menthol, (-)-menthol was significantly more potent toward 5-HT3 A- vs 5-HT3 AB receptors. We show co-expression of 5-HT3A and 5-HT3B subunits in the human gut epithelium, the lamina propria, the myenteric plexus, and the muscular cell layer. The demonstrated 5-HT3 inhibitory effects may be relevant for boldine's and menthol's alleviating properties on FGID and may encourage clinical studies with the compounds or the plant extracts for CINV and IBS treatment. The found receptor-discriminative properties make boldine and (-)-menthol to potentially useful tools for analyzing structural differences between these receptor subtypes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Chloroform, analytical standard
Sigma-Aldrich
1-Methylindol, ≥97%
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis