Merck
  • Association between PGC-1alpha gene polymorphisms and type 2 diabetes risk: a case-control study of an Iranian population.
Association between PGC-1alpha gene polymorphisms and type 2 diabetes risk: a case-control study of an Iranian population.
Canadian journal of diabetes (2014-10-06)
Shabnam Shokouhi, Karimeh Haghani, Parveneh Borji, Salar Bakhtiyari
ZUSAMMENFASSUNG
The peroxisome proliferator-activated receptor gamma coactivator-1alpha (PGC-1alpha) gene could play a role in the onset of type 2 diabetes mellitus. The aim of this study was to explore the possible associations among polymorphisms Gly482Ser, Thr394Thr and Thr528Thr of the PGC-1alpha gene and the risk of type 2 diabetes in Kurdish-Iranians. DNA specimens from all 173 type 2 diabetes subjects and 173 normoglycemic subjects were genotyped by the polymerase chain reaction-restriction fragment length polymorphism method. Genotypic and allelic frequencies were analyzed in each group. Serum lipids, fasting glucose, fasting serum insulin, homeostasis model assessment of insulin resistance and glycated hemoglobin levels were determined using the conventional methods. The data were analyzed using SPSS software. The GA genotype of Gly482Ser was associated with a significant susceptibility for type 2 diabetes (odds ratio 5.23, p<0.000). Furthermore, the GA genotype of Thr528Thr had a higher risk for type 2 diabetes (odds ratio 2.37, p<0.002). Normoglycemic persons carrying the GA+AA genotypes of Gly482Ser variation had significantly lower high-density lipoprotein cholesterol in comparison with persons having GG genotype. In comparison with GG genotype carriers, normoglycemic subjects carrying the GA+AA genotypes of Thr394Thr variation had significantly higher fasting blood sugar, fasting serum insulin and homeostasis model assessment of insulin resistance. Normoglycemic subjects with the GA+AA genotypes of Thr528Thr variation had significantly higher levels of low-density lipoprotein cholesterol compared with subjects having the GG genotype. Type 2 diabetes subjects carrying the GA+AA genotypes of this polymorphism had significantly higher waist-hip ratio in comparison with the GG genotype carriers. We also found that haplotype 394-GG/482-GA/528-GG of PGC-1alpha was significantly associated with higher risk of type 2 diabetes. Our findings revealed significant associations between PGC-1alpha Gly482Ser and Thr528Thr polymorphisms and type 2 diabetes in Kurdish-Iranians.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Phthalsäuredibutylester, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ACS reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, BioXtra, ≥99.0% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumphosphat dibasisch Dodecahydrat, tested according to Ph. Eur.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Saccharose, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Phthalsäuredibutylester, PESTANAL®, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Saccharose, analytical standard, for enzymatic assay kit SCA20
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Saccharose, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Phthalsäuredibutylester, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.