Merck
  • COX-2- and endoplasmic reticulum stress-independent induction of ULBP-1 and enhancement of sensitivity to NK cell-mediated cytotoxicity by celecoxib in colon cancer cells.
COX-2- and endoplasmic reticulum stress-independent induction of ULBP-1 and enhancement of sensitivity to NK cell-mediated cytotoxicity by celecoxib in colon cancer cells.
Experimental cell research (2014-09-15)
So-Jung Kim, Ga-Hee Ha, Jae-Ho Bae, Ga Rim Kim, Cheol-Hun Son, You-Soo Park, Kwangmo Yang, Sae-Ock Oh, Sun-Hee Kim, Chi-Dug Kang
ZUSAMMENFASSUNG
In the present study, we investigated whether celecoxib could induce the expression of NKG2D ligands in clonogenic colon cancer cells, and increase their susceptibility to NK cell-mediated cell death. Celecoxib and its non-coxib analog, 2,5-dimethyl celecoxib, induced ULBP-1 and DR5 in both COX-2 negative HCT-15 cells and COX-2 positive HT-29 cells. Celecoxib increased their susceptibility to NK92 cells in both DELFIA assay and soft agar colony forming assay. The inducibility of ULBP-1 and DR5 by celecoxib was not different between CD44- and CD44+ HCT-15 cells, and CD133- and CD133+ HT-29 cells. Celecoxib increased the susceptibility of highly clonogenic CD44+ HCT-15 and CD133+ HT-29 cells to NK92 cells, at least comparable to less clonogenic CD44- HCT-15 and CD133- HT-29 cells, respectively. In addition, celecoxib induced CHOP, and thapsigargin, an inducer of ER (endoplasmic reticulum) stress, induced DR5 but not ULBP1 in HCT-15. Taken together, these findings suggest that celecoxib induces the expression of ULBP-1 as well as DR5 in clonogenic colon cancer cells via COX-2 and ER stress-independent pathways, and increases their susceptibility to NK cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Aphidicolin, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Dimethylsulfoxid, analytical standard
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
2,5-Dimethyl-Celecoxib, ≥98% (HPLC)
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland