Merck
  • Solvent vapor annealing: an efficient approach for inscribing secondary nanostructures onto electrospun fibers.
Solvent vapor annealing: an efficient approach for inscribing secondary nanostructures onto electrospun fibers.
Macromolecular rapid communications (2014-07-22)
Jianzhao Liu, Adam J P Bauer, Bingbing Li
ZUSAMMENFASSUNG
Solvent vapor annealing (SVA) is originally developed to attain equilibrium nanostructures from microphase-separated block polymer thin films. Interestingly, by carefully choosing a solvent vapor that can selectively mobilize the amorphous chains of a semicrystalline polymer while preserving the integrity of its crystalline structure, this study demonstrates that the SVA method can also be utilized to introduce hierarchical structures onto semicrystalline polymer-based materials. This study on electrospun poly(ε-caprolactone) (PCL) fibers clearly shows that acetone, a poor solvent for PCL, can effectively delocalize the amorphous chains and redeposit them onto the pre-existing crystal edges, giving rise to secondary nanostructures inscribed onto the PCL fibers. In the past decade, various fiber fabrication methods and numerous fiber products are reported. The easy one-step approach reported here provides new insight into the design and fabrication of structurally hierarchical polymeric materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Aceton, analytical standard
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, natural, ≥97%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material