Merck
Synthesis, GluN2B affinity and selectivity of benzo[7]annulen-7-amines.
Bioorganic & medicinal chemistry (2014-12-03)
Sandeep Gawaskar, Dirk Schepmann, Alessandro Bonifazi, Bernhard Wünsch
ZUSAMMENFASSUNG
Due to their beneficial side effect profile, NMDA receptor antagonists interacting selectively with the allosteric ifenprodil binding site of the GluN2B subunit are of major interest for the treatment of neurological and neurodegenerative disorders. A series of benzo[7]annulen-7-amines 6 was designed by conformational restriction of ifenprodil (1). At first the benzo[7]annulen-7-one 11 was prepared in a three-step synthesis comprising of a double Knoevenagel condensation of phthalaldehyde (7) with dimethyl 3-oxoglutarate (8), hydrogenation of 9 and saponification/decarboxylation of 10. Reductive amination of the ketone 11 with primary amines and NaBH(OAc)3 led to the secondary amines 6a-d, cis-6h and trans-6i. The tertiary amines 6e-g were obtained by SN2-substitution of the nosylate 13. Although H-bond forming substituents in 2- and 5-position are missing, the amines 6 exhibit high affinity towards GluN2B containing NMDA receptors. A distance of four to five bond lengths between the basic amino moiety and the phenyl ring in the side chain appears to be optimal for high GluN2B affinity. The phenylcyclohexylamine cis-6h and the 4-benzylpiperidine 6g show the highest GluN2B affinities (Ki=2.3nM and 2.9nM, respectively). With respect to selectivity against the PCP binding site, σ1 and σ2 receptors the phenylpiperazine 6f is the most promising GluN2B antagonist.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tetramethylsilan, NMR grade, ACS reagent, ≥99.9%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Tetramethylsilan, ≥99.0% (GC)
Supelco
Chloroform, analytical standard
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Tetramethylsilan, analytical standard, suitable for NMR (spectroscopy), ACS reagent
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetramethylsilan, electronic grade, ≥99.99% trace metals basis