Merck
  • Multi-constituent synergism is responsible for anti-inflammatory effect of Azadirachta indica leaf extract.
Multi-constituent synergism is responsible for anti-inflammatory effect of Azadirachta indica leaf extract.
Pharmaceutical biology (2014-07-16)
Muhammad Ihtisham Umar, Mohd Zaini Asmawi, Amirin Sadikun, A M S Abdul Majid, Item Justin Atangwho, Mohamed B Khadeer Ahamed, Rabia Altaf, Ashfaq Ahmad
ZUSAMMENFASSUNG
Azadirachta indica A. Juss. (Meliaceaes) leaves have been used traditionally to treat swelling and rheumatism in Indian cultures. To fractionate A. indica leaf extracts using bioactivity guided manner for identification of the active anti-inflammatory principles. Polarity-gradient sequential extracts (petroleum ether, chloroform, methanol, and water) of A. indica leaves were screened for their anti-inflammatory potential using the carrageenan-induced rat paw edema model (1 g/kg). The chloroform extract was sequentially fractionated to obtain n-hexane (F-1), n-hexane-chloroform (F-2), and chloroform (F-3) fractions and their inhibitory effect on rat paw edema was evaluated (500 mg/kg). Inhibitory effect of F-2 on granuloma formation, plasma interleukin (IL-1), and tumor necrosis factor (TNF-α) was assessed at the doses of 100, 200, and 400 mg/kg using the cotton pellet assay in rats. Three sub-fractions (SF-1, SF-2, and SF-3) were obtained upon chromatography of F-2, and their inhibitory effect on cyclooxygenase was assessed at 200 µg/mL concentration. The sub-fractions were subjected to gas chromatography-mass spectrometry (GC-MS). All the extracts showed significant anti-inflammatory effect; however, chloroform extract was the most effective against paw edema (53.25% inhibition). The three fractions of chloroform extract showed significant effect, while F-2 being the most potent (51.02%). F-2 demonstrated dose-dependent inhibition of granuloma and cytokines. Interestingly, all the sub-fractions of F-2 inhibited COX-1 and COX-2 with almost equal potential. GC-MS revealed that chemically the sub-fractions were totally different from each other. Anti-inflammatory effect of A. indica is a result of cumulative and synergistic effects of diversified constituents with varying polarities that collectively exert the effect via suppression of cyclo-oxygenases and cytokines (IL-1 and TNF-α).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Zirkonium(IV)-oxychlorid Octahydrat, reagent grade, 98%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Methanol, analytical standard
Supelco
Hexan, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material