Merck
  • ERK-mediated phosphorylation of TFAM downregulates mitochondrial transcription: implications for Parkinson's disease.
ERK-mediated phosphorylation of TFAM downregulates mitochondrial transcription: implications for Parkinson's disease.
Mitochondrion (2014-04-29)
Kent Z Q Wang, Jianhui Zhu, Ruben K Dagda, Guy Uechi, Salvatore J Cherra, Aaron M Gusdon, Manimalha Balasubramani, Charleen T Chu
ZUSAMMENFASSUNG
Mitochondrial transcription factor A (TFAM) regulates mitochondrial biogenesis, which is downregulated by extracellular signal-regulated protein kinases (ERK1/2) in cells treated chronically with the complex I inhibitor 1-methyl-4-phenylpyridinium (MPP+). We utilized mass spectrometry to identify ERK1/2-dependent TFAM phosphorylation sites. Mutation of TFAM at serine 177 to mimic phosphorylation recapitulated the effects of MPP+ in decreasing the binding of TFAM to the light strand promoter, suppressing mitochondrial transcription. Mutant TFAM was unable to affect respiratory function or rescue the effects of MPP+ on respiratory complexes. These data disclose a novel mechanism by which ERK1/2 regulates mitochondrial function through direct phosphorylation of TFAM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Jodacetamid, BioUltra
Supelco
Tris(2-carboxyethyl)phosphin -hydrochlorid -Lösung, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Anti-HA-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
SAFC
Jodacetamid
Sigma-Aldrich
Tris(2-carboxyethyl)phosphin -hydrochlorid, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
1,3-Butadien -Lösung, 20 wt. % in toluene
SAFC
Isopropyl β-D-1-Thiogalactopyranosid
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,3-Butadien -Lösung, 15 wt. % in hexane