Merck
  • Potential of novel phytoecdysteroids isolated from Vitex doniana in the treatment depression: involvement of monoaminergic systems.
Potential of novel phytoecdysteroids isolated from Vitex doniana in the treatment depression: involvement of monoaminergic systems.
Pharmacology, biochemistry, and behavior (2014-12-03)
Ismail O Ishola, Charles O Ochieng, Sunday O Olayemi, Memunat O Jimoh, Saheed M Lawal
ZUSAMMENFASSUNG
Vitex doniana Sweet (Verbanaceae) is used in traditional African medicine for the treatment of neurological disorders including depression. In our previous studies, three new phytoecdysteroids were isolated from methanol stem bark extract of V. doniana (VD) (11β-hydroxy-20-deoxyshidasterone, 21-hydroxyshidasterone, and 2,3-acetonide-24-hydroxyecdysone) along with known ecdysteroids. This study was designed to investigate antidepressant-like effect of VD and the isolated phytoecdysteroids in behavioral models of despair, forced-swim test (FST) and tail-suspension test (TST) in mice. VD (100 and 200mg/kg, p.o.) treatment reduced (P<0.05) the duration of immobility in both tests without affecting the locomotor activity and exploratory behavior as observed in the open field test. Similarly, 21-hydroxyshidasterone, 11β-hydroxy-20-deoxyshidasterone, ajugasterone and 24-hydroxyecdysone acute oral treatments significantly reduced immobility time with peak effect at 10mg/kg, which was similar to the effect of conventional antidepressants (imipramine and fluoxetine) in the FST. Conversely, pretreatment of mice with yohimbine (1mg/kg, i.p., α2-adrenoceptor antagonist), ketanserin (5mg/kg, i.p., 5-HT2A/2C receptor antagonist) or sulpiride (dopamine D2 receptor antagonist) prevented the antidepressant-like effect of 21-hydroxyshidasterone while the effects of 11β-hydroxy-20-deoxyshidasterone and 24-hydroxyecdysone were blocked by yohimbine or ketanserin in the FST. Moreover, the anti-immobility effect elicited by ajugasterone was prevented by prazosin (62.5μg/kg, i.p., α1-adrenoceptor antagonist) pretreatment. Our findings demonstrated that V. doniana and its phytoecdysteroids constituents elicited antidepressant-like effect in behavioral paradigm of despair. Furthermore, 21-hydroxyshidasterone produces its antidepressant-like effect through interaction with α2-adrenoceptor, 5-HT2A/2C receptor and dopamine D2-receptors but 11β-hydroxy-20-deoxyshidasterone and 24-hydroxyecdysone effects depend on interaction with α2-adrenoceptor and 5-HT2A/2C receptors while ajugasterone produces its action through interaction with post-synaptic α1-adrenoceptors. Thus, phytoecdysteroids could play a pivotal role in the treatment of major depression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Prazosin -hydrochlorid, ≥99.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Yohimbin -hydrochlorid, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylsulfoxid, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.