Merck
  • Functional characterization and categorization of missense mutations that cause methylmalonyl-CoA mutase (MUT) deficiency.
Functional characterization and categorization of missense mutations that cause methylmalonyl-CoA mutase (MUT) deficiency.
Human mutation (2014-08-16)
Patrick Forny, D Sean Froese, Terttu Suormala, Wyatt W Yue, Matthias R Baumgartner
ZUSAMMENFASSUNG
Methylmalonyl-CoA mutase (MUT) is an essential enzyme in propionate catabolism that requires adenosylcobalamin as a cofactor. Almost 250 inherited mutations in the MUT gene are known to cause the devastating disorder methylmalonic aciduria; however, the mechanism of dysfunction of these mutations, more than half of which are missense changes, has not been thoroughly investigated. Here, we examined 23 patient missense mutations covering a spectrum of exonic/structural regions, clinical phenotypes, and ethnic populations in order to determine their influence on protein stability, using two recombinant expression systems and a thermostability assay, and enzymatic function by measuring MUT activity and affinity for its cofactor and substrate. Our data stratify MUT missense mutations into categories of biochemical defects, including (1) reduced protein level due to misfolding, (2) increased thermolability, (3) impaired enzyme activity, and (4) reduced cofactor response in substrate turnover. We further demonstrate the stabilization of wild-type and thermolabile mutants by chemical chaperones in vitro and in bacterial cells. This in-depth mutation study illustrates the tools available for MUT enzyme characterization, guides future categorization of further missense mutations, and supports the development of alternative, chaperone-based therapy for patients not responding to current treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumhydroxid, ACS reagent, ≥85%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, reagent grade, 90%, flakes
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kalilauge, 45 wt. % in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, ≥85% KOH basis, pellets, white
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Kalilauge, volumetric, 8.0 M KOH (8.0N)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Natriumchlorid -Lösung, 5 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, BioXtra, ≥85% KOH basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, technical, ≥85%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropyl β-D-1-Thiogalactopyranosid, ≥99% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Isopropyl-β-D-thiogalactopyranosid -Lösung, ReadyMade IPTG solution for Blue-white screening
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.