Merck
  • Concentration and distribution of antibiotics in water-sediment system of Bosten Lake, Xinjiang.
Concentration and distribution of antibiotics in water-sediment system of Bosten Lake, Xinjiang.
Environmental science and pollution research international (2014-05-09)
Xiaoning Lei, Jianjiang Lu, Zilong Liu, Yanbin Tong, Shanman Li
ZUSAMMENFASSUNG
This study investigated the residues of four sulfonamides, four quinolones, and four tetracyclines in surface water as well as surficial sediment samples, of Bosten Lake, in Xinjiang, China. The results showed the presence of 10 out of the 12 selected antibiotics in both water and sediment. Lomefloxacin was not detected in any of the samples. Among the 12 antibiotics considered, ciprofloxacin, with median concentrations of 39.22 ng L(-1) in surface water and 76.51 μg kg(-1) in surficial sediment, was the dominant antibiotic in all samples. The sorption coefficient values presented higher sorption capacities of tetracycline and chlortetracycline than the other antibiotics. The cluster analysis revealed elevated levels of pollution in sampling sites 1, 2, and 3, which were situated in a nearby urban area and in the estuary of Kaidu River. This study demonstrates the necessity of regulating the use of antibiotics and improving the management and treatment of their release.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%