Merck
  • Lovastatin and perillyl alcohol inhibit glioma cell invasion, migration, and proliferation--impact of Ras-/Rho-prenylation.
Lovastatin and perillyl alcohol inhibit glioma cell invasion, migration, and proliferation--impact of Ras-/Rho-prenylation.
Pharmacological research (2014-12-17)
Sarah Afshordel, Beatrice Kern, Jasmin Clasohm, Hildegard König, Maike Priester, Jakob Weissenberger, Donat Kögel, Gunter P Eckert
ZUSAMMENFASSUNG
Alterations in small GTPase mediated signal transduction pathways have emerged as a central step in the molecular pathogenesis of glioblastoma (GBM), the most common malignant brain tumor in adults. Farnesylpyrophosphate (FPP) and geranylgeranylpyrophosphate (GGPP) are derived from mevalonate, whose production is catalyzed by 3-hydroxy-3-methylglutaryl-CoA (HMG-CoA) reductase. Prenylation by FPP and GGPP is required for membrane insertion and oncogenic function of Ras- and Rho-proteins, within the stimulation of the Ras-Raf-MEK-ERK pathway. A straightforward prediction from HMG-CoA reductase inhibitor studies is that statins decrease FPP and GGPP levels and diminish ERK signaling ensuring less proliferation and migration of cancer cells. Perillyl alcohol (POH), a naturally occurring monoterpene inhibits prenyltransferases and is able to inhibit cancer cell growth, but the underlying mechanism is still unclear. We here report that lovastatin (LOV) and POH impair the regulation of the mevalonate- and the Ras-Raf-MEK-ERK pathway in U87 and U343 glioblastoma cells. Both compounds affected the post-translational modification of H-Ras and Rac1. While LOV diminished the substrates of the transferase reaction that catalyze prenylation, POH inhibited the enzymes itself. Our data highlight the impact of isoprenoids for post-translational modification of small GTPases promoting proliferation, migration and invasion capabilities in glioma cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Ammoniumacetat, ACS reagent, ≥97%
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O