Merck
  • Cathepsin S controls adipocytic and osteoblastic differentiation, bone turnover, and bone microarchitecture.
Cathepsin S controls adipocytic and osteoblastic differentiation, bone turnover, and bone microarchitecture.
Bone (2014-05-02)
M Rauner, U Föger-Samwald, M F Kurz, C Brünner-Kubath, D Schamall, A Kapfenberger, P Varga, S Kudlacek, A Wutzl, H Höger, P K Zysset, G P Shi, L C Hofbauer, W Sipos, P Pietschmann
ZUSAMMENFASSUNG
Cathepsin S is a cysteine protease that controls adipocyte differentiation and has been implicated in vascular and metabolic complications of obesity. Considering the inverse relation of osteoblasts and adipocytes and their mutual precursor cell, we hypothesized that cathepsin S may also affect osteoblast differentiation and bone remodeling. Thus, the fat and bone phenotypes of young (3 months old) and aged (12 or 18 months old) cathepsin S knock-out (KO) and wild-type (WT) mice were determined. Cathepsin S KO mice had a normal body weight at both ages investigated, even though the amount of subscapular and gonadal fat pads was reduced by 20%. Further, cathepsin S deficiency impaired adipocyte formation (-38%, p<0.001), which was accompanied by a lower expression of adipocyte-related genes and a reduction in serum leptin, IL-6 and CCL2 (p<0.001). Micro-CT analysis revealed an unchanged trabecular bone volume fraction and density, while tissue mineral density was significantly lower in cathepsin S KO mice at both ages. Aged KO mice further had a lower cortical bone mass (-2.3%, p<0.05). At the microarchitectural level, cathepsin S KO mice had thinner trabeculae (-8.3%), but a better connected trabecular network (+24%). Serum levels of the bone formation marker type 1 procollagen amino-terminal-propeptide and osteocalcin were both 2-3-fold higher in cathepsin S KO mice as was the mineralized surface. Consistently, osteogenic differentiation was increased 2-fold along with an increased expression of osteoblast-specific genes. Interestingly, serum levels of C-terminal telopeptide of type I collagen were also higher (+43%) in cathepsin S KO mice as were histological osteoclast parameters and ex vivo osteoclast differentiation. Thus, cathepsin S deficiency alters the balance between adipocyte and osteoblast differentiation, increases bone turnover, and changes bone microarchitecture. Therefore, bone and fat metabolisms should be monitored when using cathepsin S inhibitors clinically.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
Methanol, analytical standard